Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
med28
Entrez Gene ID
100125095
Gene Name
mediator complex subunit 28
Gene Aliases
-
Chromosome Location
Chr.1: 16439238 - 16442155 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103006.1 NP_001096476.1 2 - 3 240 101
GenBank mRNA BC136218.1 - 2 - 3 240 101
BC155038.1 - 2 - 3 220 101
CT010556.2 - 2 - 3 232 101

View Full Product Details