Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tyr
Entrez Gene ID
100125152
Gene Name
tyrosinase
Gene Aliases
-
Chromosome Location
Chr.2: 164549096 - 164562434 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103048.1 NP_001096518.1 3 - 4 1231 90
GenBank mRNA BC135591.1 - 3 - 4 1231 90
CT025512.2 - 3 - 4 1230 90

View Full Product Details