Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mad2l1
Entrez Gene ID
100125170
Gene Name
mitotic arrest deficient 2 like 1
Gene Aliases
XMad2, mad2
Chromosome Location
Chr.1: 61631061 - 61636864 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103058.1 NP_001096528.1 4 - 5 488 92
GenBank mRNA BC135782.1 - 4 - 5 488 92
CT025283.2 - 4 - 5 444 92

View Full Product Details