Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tmed2
Entrez Gene ID
734103
Gene Name
transmembrane p24 trafficking protein 2
Gene Aliases
Xp24b1, Xp24beta1, p24a, rnp24
Chromosome Location
Chr.1: 152376436 - 152383348 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045776.1 NP_001039241.1 2 - 3 387 82
GenBank mRNA CR942760.2 - 2 - 3 387 82

View Full Product Details