Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
bcl7b
Entrez Gene ID
100125186
Gene Name
B-cell CLL/lymphoma 7B
Gene Aliases
-
Chromosome Location
Chr.2: 31778838 - 31789848 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103072.1 NP_001096542.1 1 - 2 110 82
GenBank mRNA BC136023.1 - 1 - 2 110 82

View Full Product Details