Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dmrta2
Entrez Gene ID
100125187
Gene Name
DMRT like family A2
Gene Aliases
dmrt5
Chromosome Location
Chr.4: 85757943 - 85763008 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103073.1 NP_001096543.1 2 - 3 1033 75
XM_018092896.1 XP_017948385.1 1 - 2 585 75
GenBank mRNA BC136032.1 - 2 - 3 1033 75
HQ171197.1 - 1 - 2 488 75

View Full Product Details