Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mapk11
Entrez Gene ID
100101766
Gene Name
mitogen-activated protein kinase 11
Gene Aliases
p38-2, p38b, p38beta, p38beta2, prkm11, sapk2, sapk2b
Chromosome Location
Chr.3: 61982267 - 62021640 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105284.1 NP_001098754.1 6 - 7 497 69
GenBank mRNA BC152057.1 - 6 - 7 497 69

View Full Product Details