Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
bmp6
Entrez Gene ID
100127176
Gene Name
bone morphogenetic protein 6
Gene Aliases
vgr, vgr-1, vgr1
Chromosome Location
Chr.6: 83419274 - 83504740 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112907.1 NP_001106378.1 3 - 4 828 61
GenBank mRNA BC155509.1 - 3 - 4 828 61

View Full Product Details