Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cbln2
Entrez Gene ID
100127549
Gene Name
cerebellin 2 precursor
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016683375: 2082507 - 2087091 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112926.1 NP_001106397.1 2 - 3 537 80
GenBank mRNA BC152045.1 - 2 - 3 537 80

View Full Product Details