Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ccdc62
Entrez Gene ID
100127606
Gene Name
coiled-coil domain containing 62
Gene Aliases
aaa, tsp-ny
Chromosome Location
Chr.1: 152909540 - 152932076 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112963.1 NP_001106434.1 3 - 4 596 108
GenBank mRNA BC153720.1 - 3 - 4 596 108

View Full Product Details