Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
naa60
Entrez Gene ID
100127619
Gene Name
N(alpha)-acetyltransferase 60, NatF catalytic subunit
Gene Aliases
HAT4, NatF, nat15
Chromosome Location
Chr.9: 14393454 - 14405080 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112973.1 NP_001106444.1 5 - 6 566 88
XM_012969997.2 XP_012825451.1 4 - 5 570 88
GenBank mRNA BC153734.1 - 5 - 6 566 88

View Full Product Details