Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pglyrp2
Entrez Gene ID
100127677
Gene Name
peptidoglycan recognition protein 2
Gene Aliases
PGRP-L
Chromosome Location
Chr.3: 122579587 - 122583970 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113016.2 NP_001106487.2 3 - 4 1187 92
XM_012953712.1 XP_012809166.1 4 - 5 1298 92
GenBank mRNA EF095491.2 - 3 - 4 1187 92

View Full Product Details