Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
prdx5
Entrez Gene ID
100127718
Gene Name
peroxiredoxin 5
Gene Aliases
acr1, aoeb166, pmp20, prx-5, prx5, prxv, sbbi10
Chromosome Location
Chr.4: 39847328 - 39854018 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113054.1 NP_001106525.1 2 - 3 218 100
GenBank mRNA BC154896.1 - 2 - 3 218 100

View Full Product Details