Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
plekho2
Entrez Gene ID
100127744
Gene Name
pleckstrin homology domain containing O2
Gene Aliases
plekhq1, pp1628, pp9099
Chromosome Location
Chr.3: 117970259 - 117991632 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113081.1 NP_001106552.1 5 - 6 561 102
GenBank mRNA BC154931.1 - 5 - 6 561 102

View Full Product Details