Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
wscd2
Entrez Gene ID
100127745
Gene Name
WSC domain containing 2
Gene Aliases
-
Chromosome Location
Chr.1: 155727029 - 155767629 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113082.1 NP_001106553.1 1 - 2 653 79
GenBank mRNA BC154932.1 - 1 - 2 653 79

View Full Product Details