Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sbk1
Entrez Gene ID
100127848
Gene Name
SH3 domain binding kinase 1
Gene Aliases
sbk
Chromosome Location
Chr.7: 95473862 - 95479299 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113150.1 NP_001106621.1 2 - 3 241 65
GenBank mRNA BC155474.1 - 2 - 3 241 65

View Full Product Details