Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
c4a
Entrez Gene ID
100135012
Gene Name
complement component 4A (Rodgers blood group)
Gene Aliases
c4a2, c4a3, c4a4, c4a6, c4b, c4s, co4, cpamd2
Chromosome Location
Chr.8: 54217913 - 54263903 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113684.1 NP_001107156.1 13 - 14 1709 77
XM_012967939.2 XP_012823393.1 14 - 15 77
GenBank mRNA BC155689.1 - 13 - 14 1709 77

View Full Product Details