Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
efcab11
Entrez Gene ID
100135073
Gene Name
EF-hand calcium binding domain 11
Gene Aliases
c14orf143
Chromosome Location
Chr.8: 65434759 - 65468185 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113812.1 NP_001107284.1 3 - 4 295 102
GenBank mRNA BC155537.1 - 3 - 4 295 102

View Full Product Details