Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mrps7
Entrez Gene ID
100135237
Gene Name
mitochondrial ribosomal protein S7
Gene Aliases
-
Chromosome Location
Chr.10: 35787917 - 35791153 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113930.1 NP_001107402.1 4 - 5 484 71
XM_012971520.2 XP_012826974.1 4 - 5 514 71
GenBank mRNA BC157364.1 - 4 - 5 484 71
CU075466.1 - 4 - 5 491 71

View Full Product Details