Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
slc35f6
Entrez Gene ID
100135354
Gene Name
solute carrier family 35 member F6
Gene Aliases
ant2bp, c2orf18
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114029.1 NP_001107501.1 - 248 69
GenBank mRNA BC157655.1 - - 248 69
BC171138.1 - - 199 69
BC171142.1 - - 199 69

View Full Product Details