Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glipr2
Entrez Gene ID
100135358
Gene Name
GLI pathogenesis-related 2
Gene Aliases
gapr-1, xfeb
Chromosome Location
Chr.Un|NW_016683729: 1817 - 7061 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114032.1 NP_001107504.1 3 - 4 343 69
GenBank mRNA BC157662.1 - 3 - 4 343 69

View Full Product Details