Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pipox
Entrez Gene ID
548382
Gene Name
pipecolic acid oxidase
Gene Aliases
-
Chromosome Location
Chr.2: 25786668 - 25809293 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114213.1 NP_001107685.1 2 - 3 286 78
GenBank mRNA BC089750.1 - 2 - 3 261 78
BC158505.1 - 2 - 3 286 78

View Full Product Details