Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nr2f2
Entrez Gene ID
100038039
Gene Name
nuclear receptor subfamily 2 group F member 2
Gene Aliases
arp1, coup-tfii, coup1, couptfb, couptfii, svp40, tfcoup2, xCOUP-TFB, xcouptfb
Chromosome Location
Chr.3: 103946136 - 103959221 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114231.1 NP_001107703.1 2 - 3 1084 91
XM_012958361.2 XP_012813815.1 2 - 3 1665 91
XM_012958362.1 XP_012813816.1 2 - 3 614 91
GenBank mRNA BC157199.1 - 2 - 3 1084 91
CU025050.1 - 2 - 3 1233 91

View Full Product Details