Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
aqp8
Entrez Gene ID
100135726
Gene Name
aquaporin 8
Gene Aliases
-
Chromosome Location
Chr.9: 14035571 - 14051303 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114256.1 NP_001107728.1 5 - 6 929 81
XM_012970027.2 XP_012825481.1 5 - 6 900 81
GenBank mRNA BC158263.1 - 5 - 6 929 81

View Full Product Details