Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
arl6ip4
Entrez Gene ID
100135728
Gene Name
ADP ribosylation factor like GTPase 6 interacting protein 4
Gene Aliases
sr-25, srp25
Chromosome Location
Chr.1: 152735828 - 152743393 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114257.1 NP_001107729.1 5 - 6 799 92
XM_012954493.2 XP_012809947.1 5 - 6 1288 92
GenBank mRNA BC158419.1 - 5 - 6 799 92
BC161359.1 - 5 - 6 799 92

View Full Product Details