Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
emsy
Entrez Gene ID
100136947
Gene Name
EMSY, BRCA2-interacting transcriptional repressor
Gene Aliases
c11orf30, gl002
Chromosome Location
Chr.2: 168814651 - 168834152 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004912217.3 XP_004912274.1 4 - 5 735 65
XM_004912218.3 XP_004912275.1 4 - 5 734 65
XM_004912219.3 XP_004912276.1 4 - 5 735 65
XM_004912220.3 XP_004912277.1 4 - 5 734 65
GenBank mRNA BC135733.1 - 4 - 5 349 65
CT027941.2 - 3 - 4 273 65

View Full Product Details