Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
c2orf42
Entrez Gene ID
100127554
Gene Name
chromosome 2 open reading frame 42
Gene Aliases
-
Chromosome Location
Chr.3: 112156398 - 112172118 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122809.1 NP_001116281.1 7 - 8 1532 75
XM_012958482.2 XP_012813936.1 8 - 9 1891 75
XM_012958483.2 XP_012813937.1 9 - 10 2032 75
XM_012958484.2 XP_012813938.1 8 - 9 1716 75
XM_012958485.2 XP_012813939.1 8 - 9 1173 75
XM_012958486.2 XP_012813940.1 9 - 10 1311 75
GenBank mRNA BC152053.1 - 8 - 9 947 75
BC161111.1 - 7 - 8 1532 75

View Full Product Details