Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cd151
Entrez Gene ID
100144686
Gene Name
CD151 molecule (Raph blood group)
Gene Aliases
gp27, mer2, peta-3, raph, sfa1, tspan24
Chromosome Location
Chr.4: 35084908 - 35112964 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123447.1 NP_001116919.1 4 - 5 454 65
XM_012960585.2 XP_012816039.1 5 - 6 650 65
GenBank mRNA BC161493.1 - 4 - 5 454 65

View Full Product Details