Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
march5
Entrez Gene ID
100144728
Gene Name
membrane associated ring-CH-type finger 5
Gene Aliases
march-v, mitol, rnf153
Chromosome Location
Chr.7: 17394954 - 17440239 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123478.1 NP_001116950.1 4 - 5 732 90
GenBank mRNA BC160533.1 - 4 - 5 732 90

View Full Product Details