Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
c19orf54
Entrez Gene ID
100144997
Gene Name
chromosome 19 open reading frame 54
Gene Aliases
-
Chromosome Location
Chr.8: 57301580 - 57318062 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126559.1 NP_001120031.1 5 - 6 583 80
XM_012967985.1 XP_012823439.1 5 - 6 657 80
XM_012967986.2 XP_012823440.1 5 - 6 646 80
XM_012967987.2 XP_012823441.1 5 - 6 688 80
XM_012967988.2 XP_012823442.1 5 - 6 652 80
XM_012967989.2 XP_012823443.1 5 - 6 621 80
XM_018096154.1 XP_017951643.1 5 - 6 617 80
GenBank mRNA BC158337.1 - 5 - 6 583 80

View Full Product Details