Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
shpk
Entrez Gene ID
100145087
Gene Name
sedoheptulokinase
Gene Aliases
-
Chromosome Location
Chr.2: 31902486 - 31915896 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126606.1 NP_001120078.1 4 - 5 642 101
XM_012956440.1 XP_012811894.1 4 - 5 567 101
XM_012956441.2 XP_012811895.1 4 - 5 562 101
XM_018091083.1 XP_017946572.1 4 - 5 563 101
GenBank mRNA BC158517.1 - 4 - 5 642 101

View Full Product Details