Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
wnt7b
Entrez Gene ID
100145124
Gene Name
Wnt family member 7B
Gene Aliases
Xwnt-7B, Xwnt7B, wnt-7b
Chromosome Location
Chr.Un|NW_016683605: 56620 - 61593 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126633.1 NP_001120105.1 - 366 98
GenBank mRNA BC158950.1 - - 366 98

View Full Product Details