Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
rsl24d1
Entrez Gene ID
100145176
Gene Name
ribosomal L24 domain containing 1
Gene Aliases
rlp24, rpl24, rpl24l, rvas3
Chromosome Location
Chr.3: 90811794 - 90818252 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126668.1 NP_001120140.1 4 - 5 350 97
GenBank mRNA BC159094.1 - 4 - 5 350 97
CT025434.2 - 4 - 5 333 97

View Full Product Details