Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
arf3
Entrez Gene ID
100145191
Gene Name
ADP ribosylation factor 3
Gene Aliases
-
Chromosome Location
Chr.2: 133013134 - 133030828 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126681.1 NP_001120153.1 2 - 3 255 93
XM_012956457.2 XP_012811911.1 2 - 3 288 93
GenBank mRNA BC159149.1 - 2 - 3 255 93

View Full Product Details