Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
plekha3
Entrez Gene ID
100145244
Gene Name
pleckstrin homology domain containing A3
Gene Aliases
-
Chromosome Location
Chr.9: 58965054 - 58982430 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126728.1 NP_001120200.1 4 - 5 595 68
GenBank mRNA BC159323.1 - 4 - 5 595 68

View Full Product Details