Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fdx2
Entrez Gene ID
100145258
Gene Name
ferredoxin 1-like
Gene Aliases
fdx1l
Chromosome Location
Chr.1: 193284892 - 193292642 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126738.1 NP_001120210.1 3 - 4 369 84
GenBank mRNA BC159357.1 - 3 - 4 369 84

View Full Product Details