Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
prelid3a
Entrez Gene ID
100145262
Gene Name
PRELI domain containing 3A
Gene Aliases
hfl-eddg1, slmo1
Chromosome Location
Chr.6: 88520702 - 88577283 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126741.1 NP_001120213.1 4 - 5 657 93
XM_012964291.1 XP_012819745.1 4 - 5 439 93
XM_012964292.1 XP_012819746.1 4 - 5 344 93
XM_012964293.2 XP_012819747.1 4 - 5 340 93
XM_018094544.1 XP_017950033.1 3 - 4 592 93
GenBank mRNA BC159369.1 - 4 - 5 657 93

View Full Product Details