Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mettl15
Entrez Gene ID
100145263
Gene Name
methyltransferase like 15
Gene Aliases
mett5d1
Chromosome Location
Chr.4: 11374261 - 11489676 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126742.1 NP_001120214.1 3 - 4 390 77
GenBank mRNA BC159378.1 - 3 - 4 390 77

View Full Product Details