Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
map6
Entrez Gene ID
100145271
Gene Name
microtubule associated protein 6
Gene Aliases
STOP, mtap6, n-stop
Chromosome Location
Chr.2: 169573281 - 169616772 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126750.1 NP_001120222.1 1 - 2 863 101
XM_012956472.2 XP_012811926.1 1 - 2 2832 101
GenBank mRNA BC159130.1 - 1 - 2 863 101

View Full Product Details