Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
tmem65
Entrez Gene ID
100145439
Gene Name
transmembrane protein 65
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016683417: 270734 - 312049 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126893.1 NP_001120365.1 1 - 2 444 76
GenBank mRNA BC161055.1 - 1 - 2 444 76

View Full Product Details