Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tymp
Entrez Gene ID
100145512
Gene Name
thymidine phosphorylase
Gene Aliases
-
Chromosome Location
Chr.3: 79833778 - 79856917 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126955.1 NP_001120427.1 4 - 5 547 63
GenBank mRNA BC161182.1 - 4 - 5 547 63

View Full Product Details