Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ankdd1b
Entrez Gene ID
100145596
Gene Name
ankyrin repeat and death domain containing 1B
Gene Aliases
-
Chromosome Location
Chr.1: 165417399 - 165443858 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127008.1 NP_001120480.1 3 - 4 495 60
XM_012956326.2 XP_012811780.1 3 - 4 343 60
XM_018091063.1 XP_017946552.1 3 - 4 367 60
GenBank mRNA BC161348.1 - 3 - 4 495 60

View Full Product Details