Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sde2
Entrez Gene ID
100145674
Gene Name
SDE2 telomere maintenance homolog (S. pombe)
Gene Aliases
c1orf55
Chromosome Location
Chr.5: 4501714 - 4507578 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127056.1 NP_001120528.1 1 - 2 121 76
GenBank mRNA CR762124.2 - 1 - 2 121 76

View Full Product Details