Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
vbp1
Entrez Gene ID
100145749
Gene Name
von Hippel-Lindau binding protein 1
Gene Aliases
-
Chromosome Location
Chr.8: 29477331 - 29484786 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127120.1 NP_001120592.1 1 - 2 99 87
GenBank mRNA BC077711.1 - 1 - 2 87 87
BC161613.1 - 1 - 2 99 87
CU025132.1 - 1 - 2 76 87

View Full Product Details