The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126522.1 | NP_001119994.1 | 2 - 3 | 717 | 101 |
GenBank mRNA | BC158255.1 | - | 2 - 3 | 717 | 101 |
CU024918.1 | - | 2 - 3 | 342 | 101 |