Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
carnmt1
Entrez Gene ID
100144950
Gene Name
carnosine N-methyltransferase 1
Gene Aliases
c9orf41
Chromosome Location
Chr.1: 119231116 - 119241478 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126522.1 NP_001119994.1 4 - 5 1014 113
GenBank mRNA BC158255.1 - 4 - 5 1014 113
CU024918.1 - 4 - 5 639 113

View Full Product Details