Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ska1
Entrez Gene ID
100144953
Gene Name
spindle and kinetochore associated complex subunit 1
Gene Aliases
C18orf24
Chromosome Location
Chr.1: 191634330 - 191641768 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126525.1 NP_001119997.1 2 - 3 170 101
XM_012955600.2 XP_012811054.1 2 - 3 231 101
XM_012955603.2 XP_012811057.1 2 - 3 232 101
GenBank mRNA BC158262.1 - 2 - 3 170 101
CT025397.2 - 2 - 3 163 101

View Full Product Details