Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cpa6
Entrez Gene ID
100144967
Gene Name
carboxypeptidase A6
Gene Aliases
cpah
Chromosome Location
Chr.6: 104508346 - 104574241 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126533.1 NP_001120005.1 4 - 5 482 104
GenBank mRNA BC158290.1 - 4 - 5 482 104

View Full Product Details