Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cwc25
Entrez Gene ID
100145020
Gene Name
CWC25 spliceosome-associated protein homolog
Gene Aliases
ccdc49
Chromosome Location
Chr.10: 3809578 - 3821372 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126572.2 NP_001120044.1 8 - 9 945 78
GenBank mRNA BC158381.1 - 8 - 9 947 78

View Full Product Details