Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cln6
Entrez Gene ID
100145280
Gene Name
CLN6, transmembrane ER protein
Gene Aliases
-
Chromosome Location
Chr.3: 116440479 - 116449427 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126758.1 NP_001120230.1 4 - 5 565 98
GenBank mRNA BC160405.1 - 4 - 5 565 98

View Full Product Details